missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CLASP2 (aa 875-914) Control Fragment Recombinant Protein
Klik for at se tilgængelige muligheder
Quantity:
100 μL
Pakningsstørrelse:
100µL
Beskrivelse
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66893 (PA5-66893. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CLASP2 is microtubule plus-end tracking protein that promotes the stabilization of dynamic microtubules. Required for the polarization of the cytoplasmic microtubule arrays in migrating cells towards the leading edge of the cell. May act at the cell cortex to enhance the frequency of rescue of depolymerizing microtubules by attaching their plus-ends to cortical platforms composed of ERC1 and PHLDB2. This cortical microtubule stabilizing activity is regulated at least in part by phosphatidylinositol 3-kinase signaling. Also performs a similar stabilizing function at the kinetochore which is essential for the bipolar alignment of chromosomes on the mitotic spindle.
Tekniske data
Tekniske data
| Accession Number | O75122 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23122 |
| Name | Human CLASP2 (aa 875-914) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1500004F14Rik; 8030404L10Rik; C77448; CLASP2; CLASP2alpha; CLASP2beta; CLASP2gamma; CLIP associating protein 2; CLIP-associating protein 2; CLIP-associating protein CLASP2; cytoplasmic linker associated protein 2; cytoplasmic linker-associated protein 2; hOrbit2; Kiaa0627; mKIAA0627; multiple asters (Mast)-like homolog 2; protein Orbit homolog 2 |
| Common Name | CLASP2 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion