missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CL-P1/COLEC12 (NP_569057) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | CL-P1/COLEC12 |
| Accession Number | NP_569057 |
| Gene ID (Entrez) | 81035 |
| Gene Alias | CL-P1, CLP1hCL-P1, COLEC12, collectin placenta 1, Collectin placenta protein 1, collectin sub-family member 12, collectin-12, NSR2, Nurse cell scavenger receptor 2, SCARA4, SCARA4NSR2, Scavenger receptor class A member 4, scavenger receptor class A, member 4, SRCL Type I, SRCLScavenger receptor with C-type lectin |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction