missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human cIAP-1/HIAP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB818-SP
This item is not returnable.
View return policy
Description
cIAP-1/HIAP-2 Monoclonal specifically detects cIAP-1/HIAP-2 in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| cIAP-1/HIAP-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Knockout Validated | |
| Q13490 | |
| BIRC2 | |
| E. coli-derived recombinant human cIAP-1/HIAP-2 His2-Ser618 Accession # Q13490 | |
| 25 μg | |
| Primary | |
| Detects human cIAP-1/HIAP-2 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) cIAP-2 (aa 2-604) and no cross-reactivity with rhBIRC6 (aa 4582-4735), rhcIAP-2 (aa 94-178), rhXIAP (aa 1-497), rhXIAP (BIR2 domain; aa 124-242), or rhXIAP (BIR3 domain; aa 252-356) is observed. | |
| Human | |
| Purified |
| Western Blot, KnockDown | |
| 681732 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, BIRC2, cIAP1, C-IAP1, hIAP2, HIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHB, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 329 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction