Learn More
Human CHRM5 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB10323V-100UG
Product Code. 18794918
  • £441.00 / 100µg
Description
Muscarinic Acetylcholine Receptor M5/CHRM5 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M5/CHRM5 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Muscarinic Acetylcholine Receptor M5/CHRM5
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
cholinergic receptor, muscarinic 5, HM5, M5, M5R, mAChR M5, MGC41838, muscarinic acetylcholine receptor M5
NS0 mouse myeloma cell line transfected with human CHRM5, Met1-Pro532, Accession # P08912
100 μg
Primary
Human
Purified
Flow Cytometry
590123
Flow Cytometry 0.25-1 ug/10^6 cells
P08912
Mouse
Protein A or G purified from ascites
RUO
1133
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2a