missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ChREBP (aa 608-658) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84659 (PA5-84659. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.
Specifications
Specifications
| Accession Number | Q9NP71 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51085 |
| Name | Human ChREBP (aa 608-658) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | BHLHD14; carbohydrate response element binding protein; carbohydrate-responsive element-binding protein; ChREBP; Class D basic helix-loop-helix protein 14; MIO; MLX interacting protein like; MLX interacting protein-like; MLX interacting protein-like beta; Mlx interactor; MLX-interacting protein-like; MLXIPL; MONDOB; putative hepatic transcription factor; WBSCR14; Williams Beuren syndrome chromosome region 14; williams-Beuren syndrome chromosomal region 14 protein; Williams-Beuren syndrome chromosomal region 14 protein homolog; Williams-Beuren syndrome chromosome region 14 homolog; Williams-Beuren syndrome chromosome region 14 protein 1; Williams-Beuren syndrome chromosome region 14 protein 2; Williams-Beuren syndrome chromosome region 14 protein 3; WS basic-helix-loop-helix leucine zipper protein; WS-bHLH |
| Common Name | ChREBP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction