missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CHRDL1 Recombinant Protein, PeproTech®

Product Code. 30245156
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 30245156

Brand: Gibco™ 45067250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

CHRDL1 is an antagonist of bone morphogenetic protein 4. The encoded protein may play a role in topographic retinotectal projection and in the regulation of retinal angiogenesis in response to hypoxia. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BU40
For Use With (Application) Functional Assay
Formulation Protein with no preservative
Gene ID (Entrez) 91851
Molecular Weight (g/mol) 48.8 kDa
Name CHRDL1
Quantity 250 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias CHL; CHL1; chordin like 1; chordin-like 1; chordin-like protein 1; Chrdl1; dA141H5.1; Kjr; Kohjirin; MGC1; MGCN; neuralin 1; neuralin-1; neurogenesin 1; Neurogenesin-1; Ng1; Nrln1; Ventroptin; ventroptin-alpha; ventroptin-beta; VOPT
Gene Symbol CHRDL1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence EQVKHSETYC MFQDKKYRVG ERWHPYLEPY GLVYCVNCIC SENGNVLCSR VRCPNVHCLS PVHIPHLCCP RCPDSLPPVN NKVTSKSCEY NGTTYQHGEL FVAEGLFQNR QPNQCTQCSC SEGNVYCGLK TCPKLTCAFP VSVPDSCCRV CRGDGELSWE HSDGDIFRQP ANREARHSYH RSHYDPPPSR QAGGLSRFPG ARSHRGALMD SQQASGTIVQ IVINNKHKHG QVCVSNGKTY SHGESWHPNL RAFGIVECVL CTCNVTKQEC KKIHCPNRYP CKYPQKIDGK CCKVCPGKKA KELPGQSFDN KGYFCGEETM PVYESVFMED GETTRKIALE TERPPQVEVH VWTIRKGILQ HFHIEKISKR MFEELPHFKL VTRTTLSQWK IFTEGEAQIS QMCSSRVCRT ELEDLVKVLY LERSEKGHC
Activity Determined by its ability to induce alkaline phosphatase production by ADTC-5 cells.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.