missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chorionic Gonadotropin alpha/beta (HCG) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£167.00 - £428.00
Specifications
| Antigen | hCG |
|---|---|
| Clone | 1270D |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713697
![]() |
R&D Systems
MAB97702-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18029221
![]() |
R&D Systems
MAB97702-100 |
100 μg |
£428.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
hCG Monoclonal specifically detects hCG in Human samples. It is validated for ELISA.Specifications
| hCG | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P0DN86 nad P0DN87 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 1270D | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human embryonic kidney cell HEK293-derived transfected with human Chorionic Gonadotropin beta , Accession # P0DN86 nad P0DN87 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title