missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chordin-like 1/CHRDL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1808
This item is not returnable.
View return policy
Description
Chordin-like 1/CHRDL1 Polyclonal specifically detects Chordin-like 1/CHRDL1 in Human samples. It is validated for Western Blot.
Specifications
| Chordin-like 1/CHRDL1 | |
| Polyclonal | |
| Unconjugated | |
| NP_001137454 | |
| CHRDL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Chordin-like 1/CHRDL1 Glu28-Cys457 Accession # NP_001137454 | |
| 100 μg | |
| Primary | |
| Detects human Chordin-like 1/CHRDL1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CHL, chordin-like 1, chordin-like protein 1, CHRDL1, CRDL1, dA141H5.1, Neuralin 1, neuralin-1, Neurogenesin-1, NRLN1, NRLN1neuralin 1, ventroptin, Ventroptin alpha, VOPT | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 91851 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction