missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chordin-like 1/CHRDL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £369.00
Specifications
| Antigen | Chordin-like 1/CHRDL1 |
|---|---|
| Clone | 263737 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772418
![]() |
R&D Systems
MAB1808-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18132801
|
R&D Systems
MAB1808 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chordin-like 1/CHRDL1 Monoclonal specifically detects Chordin-like 1/CHRDL1 in Human samples. It is validated for Western Blot.Specifications
| Chordin-like 1/CHRDL1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_001137454 | |
| 91851 | |
| Mouse myeloma cell line NS0-derived recombinant human Chordin-like 1/CHRDL1 Glu28-Cys457 Accession # NP_001137454 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Chordin-like 1/CHRDL1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Chordin-Like 2 is observed. |
| 263737 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHL, chordin-like 1, chordin-like protein 1, CHRDL1, CRDL1, dA141H5.1, Neuralin 1, neuralin-1, Neurogenesin-1, NRLN1, NRLN1neuralin 1, ventroptin, Ventroptin alpha, VOPT | |
| CHRDL1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title