missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CHMP2B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7509-SP
This item is not returnable.
View return policy
Description
CHMP2B Monoclonal specifically detects CHMP2B in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CHMP2B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q9UQN3 | |
| CHMP2B | |
| E. coli-derived recombinant human CHMP2B Ala2-Asp213 Accession # Q9UQN3 | |
| 25 μg | |
| Primary | |
| Detects human CHMP2B in direct ELISAs and Western blots | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 791521 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| charged multivesicular body protein 2b, CHMP2.5, CHMP2.5VPS2BVPS2-2, CHMP2b, chromatin modifying protein 2B, Chromatin-modifying protein 2b, DKFZP564O123, DMT1, hVps2-2, Vacuolar protein sorting-associated protein 2-2, vacuolar protein-sorting-associated protein 2-2, VPS2 homolog B, Vps2-2, VPS2B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25978 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction