missing translation for 'onlineSavingsMsg'
Learn More

Human CHL-1/L1CAM-2 Antibody, R&D Systems™
GREENER_CHOICE

Goat Polyclonal Antibody

£168.00 - £452.00

Specifications

Antigen CHL-1/L1CAM-2
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18759097
GREENER_CHOICE
View Documents
R&D Systems
AF2126-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18089621
GREENER_CHOICE
View Documents
R&D Systems
AF2126
100 μg
£452.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CHL-1/L1CAM-2 Polyclonal specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CHL-1/L1CAM-2
Western Blot 0.1 ug/mL
Polyclonal
Goat
Human
CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein
CHL1
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Western Blot
Unconjugated
RUO
EAW63869
10752
Mouse myeloma cell line NS0-derived recombinant human CHL-1/L1CAM-2 Ile25-Gln1096 Accession # EAW63869
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects human CHL-1/L1CAM-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse CHL-1 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.