missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chk2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1358
This item is not returnable.
View return policy
Description
Chk2 Polyclonal specifically detects Chk2 in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| Chk2 | |
| Polyclonal | |
| Unconjugated | |
| O96017 | |
| CHEK2 | |
| E. coli-derived recombinant human Chk2 Asp208-Leu543 Accession # O96017 | |
| 100 μg | |
| Primary | |
| Detects human Chk2 in Western blots. | |
| Human | |
| IgG |
| Western Blot, KnockDown | |
| LYOPH | |
| Western Blot 1 ug/mL, Knockout Validated | |
| CDS1, CHEK2, CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, HuCds1, LFS2, PP1425, Rad53, S.pombe) homolog | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11200 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction