missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CHCHD7 Synthetic Peptide

Product Code. 15295449
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 15295449

Brand: Invitrogen™ PEP1529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

CHCHD7 Synthetic Peptide, PEP-1529, from Invitrogen.

CHCHD7 is a newly identified member of a multifamily of proteins containing a conserved (coiled coil 1)-(helix 1)-(coiled coil 2)-(helix 2) domain. Northern blot analysis revealed that the gene is ubiquitously expressed. Its biological function is unknown and the gene has hitherto not been associated with neoplasia. CHCHD7 and PLAG1 are located head-to-head about 500 bp apart in 8q12. A chromosomal aberration involving CHCHD7 is found in salivary gland pleiomorphic adenomas, the most common benign epithelial tumors of the salivary gland.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 79145
Species Human
Purification Method purified
Accession Number Q9BUK0
Common Name Human CHCHD7
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol CHCHD7
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.