missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human cGKI (aa 233-349) Control Fragment Recombinant Protein

Product Code. 30211306
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211306

Brand: Invitrogen™ RP89820

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82310 (PA5-82310. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PRKG1 is a cGMP-dependent serine/threonine kinase which has an important role in the relaxation of vascular smooth muscle and the inhibition of platelet aggregation. The PRKG1 proteins play a central role in regulating cardiovascular and neuronal functions in addition to relaxing smooth muscle tone, and modulating cell growth. This gene is most strongly expressed in all types of smooth muscle, platelets, cerebellar Purkinje cells, hippocampal neurons, and the lateral amygdala. Isoforms Ialpha and Ibeta have identical cGMP-binding and catalytic domains but differ in their leucine/isoleucine zipper and autoinhibitory sequences and therefore differ in their dimerization substrates and kinase enzyme activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13976
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5592
Name Human cGKI (aa 233-349) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AAT8; AW125416; cGK; cGK 1; CGK 1 alpha; cGK 1 beta; cGK cGK cGKI; cGK cGK1; cGK1; cGKI; cGKI-alpha; cGKI-BETA; cGMP dependent protein kinase type 1; cGMP kinase type I alpha; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase 1, alpha isozyme; cGMP-dependent protein kinase 1, beta isozyme; cGMP-dependent protein kinase I; Gm19690; OTTHUMP00000019618; Pk.; Pkgi; Prkg1; PRKG1B; PRKGR1A; PRKGR1B; protein kinase cGMP-dependent 1; protein kinase, cGMP-dependent, regulatory, type I, beta; protein kinase, cGMP-dependent, type 1; protein kinase, cGMP-dependent, type I; RP11-346D6.1; Unknown (protein for MGC:134126); unnamed protein product
Common Name cGKI
Gene Symbol PRKG1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LADVLEETHYENGEYIIRQGARGDTFFIISKGTVNVTREDSPSEDPVFLRTLGKGDWFGEKALQGEDVRTANVIAAEAVTCLVIDRDSFKHLIGGLDDVSNKAYEDAEAKAKYEAEA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.