missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CG beta (HCG beta) DuoSet ELISA
Description
The Human CG beta (HCG beta) DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Chorionic Gonadotropin beta Chain (hCG beta).
Specifications
Specifications
| Assay Range | 7.81 - 500pg/mL |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1082 |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?