missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CFL1 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£206.00 - £486.00
Specifications
| Antigen | Cofilin |
|---|---|
| Clone | 1093310 |
| Dilution | Western Blot 0.5 μg/mL, Simple Western 10-100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Applications | Simple Western, Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30484807
|
R&D Systems
MAB11669-100 |
100 μg |
£486.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30484804
|
R&D Systems
MAB11669-SP |
25 μg |
£206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cofilin Monoclonal antibody specifically detects Cofilin in Human samples. It is validated for Simple Western,Western Blot,Immunohistochemistry,ImmunocytochemistrySpecifications
| Cofilin | |
| Western Blot 0.5 μg/mL, Simple Western 10-100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| 18 kDa phosphoprotein, CFL, cofilin, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, HEL-S-15, p18 | |
| Synthetic Peptide, Accession # P23528 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Antibody |
| 1093310 | |
| Simple Western, Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P23528 | |
| 1072 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title