missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CES2 (aa 334-442) Control Fragment Recombinant Protein

Product Code. 30207819
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207819

Brand: Invitrogen™ RP101044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (66%), Rat (66%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82645 (PA5-82645. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CES2 is a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. The protein encoded by this gene is the major intestinal enzyme and functions in intestine drug clearance.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00748
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8824
Name Human CES2 (aa 334-442) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9030624L02Rik; 9130231C15Rik; ACH M1; Acylcarnitine hydrolase; acylcarnitine hydrolase M1; AI266984; Carboxylesterase 2; carboxylesterase 2 (intestine, liver); carboxylesterase 2 A; carboxylesterase 2 C; carboxylesterase 2 E; carboxylesterase 5; carboxylesterase 6; carboxylesterase isoenzyme; carboxylesterase ML3; CE-2; CES 2; Ces2; CES-2; Ces2a; CES2A1; ces2A3; Ces2c; Ces2e; Ces2l; Ces5; Ces6; CESRL4; Cocaine esterase; hCE-2; ICE; intestinal carboxylesterase; liver carboxylesterase-2; methylumbelliferyl-acetate deacetylase 2; P iCE; PCE-2; Peroxisome proliferator-inducible acylcarnitine hydrolase; phenobarbital-inducible carboxylesterase (liver); pyrethroid hydrolase Ces2a; Pyrethroid hydrolase Ces2e
Common Name CES2
Gene Symbol CES2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PVPSIVGVNNNEFGWLIPKVMRIYDTQKEMDREASQAALQKMLTLLMLPPTFGDLLREEYIGDNGDPQTLQAQFQEMMADSMFVIPALQVAHFQCSRAPVYFYEFQHQP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.