missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cervical Adenocarcinoma (HeLa-S3) Total RNA
Spécification
Spécification
| Species | Human |
| Sample Type | RNA |
| For Use With (Application) | Nucleic Acid Quantitation |
| Product Type | Total RNA |
| Quantity | 10 μg |
| Sample Source | Tumor or Diseased Tissue, Cell Line |
| Shipping Condition | Dry Ice |
Nom du produit
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?