missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CENPF (aa 620-761) Control Fragment Recombinant Protein

Product Code. 30206440
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206440

Brand: Invitrogen™ RP110098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144905 (PA5-144905. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein that associates with the centromere-kinetochore complex. The protein is a component of the nuclear matrix during the G2 phase of interphase. In late G2 the protein associates with the kinetochore and maintains this association through early anaphase. It localizes to the spindle midzone and the intracellular bridge in late anaphase and telophase, respectively, and is thought to be subsequently degraded. The localization of this protein suggests that it may play a role in chromosome segregation during mitosis. It is thought to form either a homodimer or heterodimer. Autoantibodies against this protein have been found in patients with cancer or graft versus host disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49454
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1063
Name Human CENPF (aa 620-761) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6530404A22Rik; AH antigen; AI325968; cell-cycle-dependent 350 K nuclear protein; CENF; CENPF; CENP-F; CENP-F kinetochore protein; centromere autoantigen F; centromere protein F; centromere protein F, (mitosin); centromere protein F, 350/400 ka (mitosin); centromere protein F, 350/400 kDa (mitosin); CILD31; hcp-1; im:7140452; im:7144238; im:7154719; im:7156761; kinetochore protein CENPF; Lek1; leucine, glutamic acid, lysine family 1 protein; mitosin; PRO1779; RP11-262H5.1; si:ch211-152h14.2; STROMS
Common Name CENPF
Gene Symbol CENPF
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SCWKSENEKLLTQMESEKENLQSKINHLETCLKTQQIKSHEYNERVRTLEMDRENLSVEIRNLHNVLDSKSVEVETQKLAYMELQQKAEFSDQKHQKEIENMCLKTSQLTGQVEDLEHKLQLLSNEIMDKDRCYQDLHAEYE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.