missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CELSR3 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7278P
This item is not returnable.
View return policy
Description
Flamingo homolog 1/CELSR3 Monoclonal specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Flow Cytometry.
Specifications
| Flamingo homolog 1/CELSR3 | |
| Monoclonal | |
| R-PE | |
| Q9NYQ7 | |
| CELSR3 | |
| E. coli-derived recombinant human CELSR3 Val531-Ser711 Accession # Q9NYQ7 | |
| 100 Tests | |
| 1951 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 763103 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11, CDHF11RESDA1, EGFL1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2, RESDA1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CELSR3 in ELISA. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction