missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CELSR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7278-SP
This item is not returnable.
View return policy
Description
Flamingo homolog 1/CELSR3 Monoclonal specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Flamingo homolog 1/CELSR3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9NYQ7 | |
| CELSR3 | |
| E. coli-derived recombinant human CELSR3 Val531-Ser711 Accession # Q9NYQ7 | |
| 25 μg | |
| Primary | |
| Detects human CELSR3 in ELISA. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 763103 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11, CDHF11RESDA1, EGFL1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2, RESDA1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1951 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction