missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CEACAM-5/CD66e (P06731) VersaClone cDNA

Product Code. 18753729
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18753729

Brand: R&D Systems™ RDC0954

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carcinoembryonic antigen-related cell adhesion molecule 5

The Human CEACAM-5/CD66e (P06731) VersaClone cDNA clone contains the complete ORF for CEACAM5/CD66e, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CEACAM5/CD66e insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Gene CEACAM5/CD66e
Accession Number P06731
Gene ID (Entrez) 1048
Gene Alias Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e, CD66e antigen, CEA, CEACAM5, CEACD66e, DKFZp781M2392, Meconium antigen 100
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.