missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-4 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7845U-100UG
This item is not returnable.
View return policy
Description
CEACAM4 Monoclonal specifically detects CEACAM4 in Human samples. It is validated for Flow Cytometry.
Specifications
| CEACAM4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O75871 | |
| CEACAM4 | |
| E. coli-derived recombinant human CEACAM-4 Phe36-Val140 Accession # O75871 | |
| RUO | |
| 1089 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 822608 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| carcinoembryonic antigen CGM7, carcinoembryonic antigen gene family member 7, carcinoembryonic antigen-related cell adhesion molecule 4, CEACAM4, CGM7, CGM7_HUMAN | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CEACAM-4 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction