missing translation for 'onlineSavingsMsg'
Learn More

Human CEACAM-3/CD66d Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7065969
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18718508 25 μg 25µg
18160921 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18718508 Supplier R&D Systems Supplier No. MAB4166SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM3/CD66d Monoclonal specifically detects CEACAM3/CD66d in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM3/CD66d
Applications Western Blot
Classification Monoclonal
Clone 417009
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P40198
Gene Alias CD66D, CEA, CEACAM3 carcinoembryonic antigen-related cell adhesion molecule 3, CGM1, W264, W282
Gene Symbols CEACAM3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CEACAM-3/CD66d isoform 1 Lys35-Gly155 Accession # P40198
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1084
Test Specificity Detects human CEACAM-3/CD66d in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CEACAM-1, -4, -5, or -6 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.