missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £417.00
Specifications
| Antigen | CEACAM19 |
|---|---|
| Clone | 1032215 |
| Dilution | Flow Cytometry 0.25 ug/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704397
![]() |
R&D Systems
MAB9397-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760547
![]() |
R&D Systems
MAB9397-100 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEACAM19 Monoclonal antibody specifically detects CEACAM19 in Human samples. It is validated for Flow CytometrySpecifications
| CEACAM19 | |
| Flow Cytometry 0.25 ug/106 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| carcinoembryonic antigen-related cell adhesion molecule 19, CEACAM19, CEAL1 | |
| Human embryonic kidney cell HEK293-derived human CEACAM-19, Ala33-Gly157, Accession # Q7692-1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1032215 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q7692-1 | |
| 56971 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title