missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-1/CD66a Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2244
This item is not returnable.
View return policy
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-reported.
Specifications
| CEACAM1/CD66a | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-reported | |
| P13688 | |
| CEACAM1 | |
| Mouse myeloma cell line NS0-derived recombinant human CEACAM-1 Gln35-Gly428 Accession # P13688 | |
| 100 μg | |
| Primary | |
| Detects human CEACAM-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) CD31, rhICAM-1, -2, -3, recombinant mouse MAdCAM-1, or rhVCAM-1. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 283340 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antigen CD66, BGP1, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, Biliary Glycoprotein 1, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen, Cea-1, CEACAM1, Hv-1, Hv-2, MHVR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 634 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction