missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDK2AP1 (aa 30-77) Control Fragment Recombinant Protein

Product Code. 30197285
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197285

Brand: Invitrogen™ RP106793

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66098 (PA5-66098. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a cyclin-dependent kinase 2 (CDK2) -associated protein which is thought to negatively regulate CDK2 activity by sequestering monomeric CDK2, and targeting CDK2 for proteolysis. This protein was found to also interact with DNA polymerase alpha/primase and mediate the phosphorylation of the large p180 subunit, which suggests a regulatory role in DNA replication during the S-phase of the cell cycle. This protein also forms a core subunit of the nucleosome remodeling and histone deacetylation (NURD) complex that epigenetically regulates embryonic stem cell differentiation. This gene thus plays a role in both cell-cycle and epigenetic regulation. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14519
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8099
Name Human CDK2AP1 (aa 30-77) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Apc10; CDK2 (cyclin-dependent kinase 2)-associated protein 1; Cdk2ap1; CDK2-associated protein 1; CDKAP1; cyclin dependent kinase 2 associated protein 1; cyclin-dependent kinase 2 associated protein 1; cyclin-dependent kinase 2-associated protein 1; Deleted in oral cancer 1; Deleted in oral cancer-1; Doc1; doc-1; DORC1; p12; p12DOC-1; Putative oral cancer suppressor; ST19
Common Name CDK2AP1
Gene Symbol CDK2AP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ATSSQYRQLLSDYGPPSLGYTQGTGNSQVPQSKYAELLAIIEELGKEI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.