missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDIP1 Control Fragment Recombinant Protein

Product Code. 30212295
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212295

Brand: Invitrogen™ RP101138

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61940 (PA5-61940. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The p53 tumor-suppressor gene integrates numerous signals that control cell life and death; loss of its functions contributes to the development of most cancers. CDIP is a novel pro-apoptotic target gene whose inhibition abrogates p53-mediated apoptotic responses. Overexpression of CDIP induced apoptosis in transfected cells while siRNA suppression of caspase-8 mRNA blocked this CDIP-induced apoptosis, indicating that the CDIP-dependent apoptosis pathway proceeds through extrinsic cell death pathway. CDIP may thus represent a novel target for drug design to maximize p53 response and sensitize tumor cells to cancer therapy. Multiple isoforms of CDIP are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H305
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29965
Name Human CDIP1 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700048O17Rik; 5730403B10Rik; AU018521; AW742692; C16orf5; CDIP; CDIP1; cell death inducing protein; cell death inducing Trp53 target 1; cell death involved p53-target; cell death-inducing p53 target 1; cell death-inducing p53-target protein 1; Cell death-inducing protein; I1; Lipopolysaccharide-induced tumor necrosis factor-alpha-like protein; LITAFL; LITAF-like protein; Transmembrane protein I1
Common Name CDIP1
Gene Symbol CDIP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CLINDFKDVTHTCPSCKAYIYTYKLLC
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.