missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDH22 (aa 281-391) Control Fragment Recombinant Protein

Product Code. 30198184
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198184

Brand: Invitrogen™ RP90328

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52919 (PA5-52919. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a member of the cadherin superfamily. The gene product is composed of five cadherin repeat domains and a cytoplasmic tail similar to the highly conserved cytoplasmic region of classical cadherins. Expressed predominantly in the brain, this putative calcium-dependent cell adhesion protein may play an important role in morphogenesis and tissue formation in neural and non-neural cells during development and maintenance of the brain and neuroendocrine organs. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UJ99
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64405
Name Human CDH22 (aa 281-391) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C20orf25; Cadherin; cadherin 22; cadherin 22, type 2; cadherin-22; cadherin-like 22; CDH22; dJ998H6.1; ortholog of rat PB-cadherin; PB-cadherin; Pituitary and brain cadherin
Common Name CDH22
Gene Symbol CDH22
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MYQFSIQESAPIGTAVGRVKAEDSDVGENTDMTYHLKDESSSGGDVFKVTTDSDTQEAIIVVQKRLDFESQPVHTVILEALNKFVDPRFADLGTFRDQAIVRVAVTDVDEP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.