missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cdc42EP5 (aa 118-148) Control Fragment Recombinant Protein

Product Code. 30180739
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180739 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30180739 Supplier Invitrogen™ Supplier No. RP99145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (58%), Rat (58%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60367 (PA5-60367. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Cell division control protein 42 (CDC42), a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg (binder of Rho GTPases) family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to CDC42 and regulate its function negatively. The encoded protein may inhibit c-Jun N-terminal kinase (JNK) independently of CDC42 binding. The protein may also play a role in septin organization and inducing pseudopodia formation in fibroblasts [provided by RefSeq, Jul 2013].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6NZY7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 148170
Name Human Cdc42EP5 (aa 118-148) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700027J19Rik; 2010007O02Rik; binder of Rho GTPase 3; binder of Rho GTPases 3; BORG3; C85526; CDC42 effector protein (Rho GTPase binding) 5; CDC42 effector protein 5; CDC42EP5; Cep5; CRIB-containing BORG3 protein
Common Name CDC42EP5
Gene Symbol CDC42EP5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KPDAEPRPGTQPPQARCRPNADLELNDVIGL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.