missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD98 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5920-SP
This item is not returnable.
View return policy
Description
CD98 Monoclonal specifically detects CD98 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| CD98 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P08195 | |
| SLC3A2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD98 Asp105-Gln529 Accession # P08195 | |
| 25 μg | |
| Primary | |
| Detects human CD98 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 590559 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, CH98hc, FRP-1, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, SLC3A2, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6520 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction