missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD97 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB25291R-SP
This item is not returnable.
View return policy
Description
CD97 Isoform 1 Monoclonal specifically detects CD97 Isoform 1 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CD97 Isoform 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD97, TM7LN1 | |
| Mouse myeloma cell line NS0-derived recombinant human CD97 Gln21-Gln398 Accession # NP_001775.2 | |
| 25 μg | |
| 976 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization, ELISA | |
| 380905R | |
| Neutralization 0.25-1.25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_001775.2 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human CD97 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction