missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CD93 Partial ORF (ABM85160.1, 24 a.a. - 580 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022918-Q03.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16075305

  • £425.00 / 25µg
Estimated Shipment: 08-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton. [provided by RefSeq]

Sequence: ADTEAVVCVGTACYTAHSGKLSAAEAQNHCNQNGGNLATVKSKEEAQHVQRVLAQLLRREAALTARMSKFWIGLQREKGKCLDPSLPLKGFSWVGGGEDTPYSNWHKELRNSCISKRCVSLLLDLSQPLLPSRLPKWSEGPCGSPGSPGSNIEGFVCKFSFKGMCRPLALGGPGQVTYTTPFQTTSSSLEAVPFASAANVACGEGDKDETQSHYFLCKEKAPDVFDWGSSGPLCVSPKYGCNFNNGGCHQDCFEGGDGSFLCGCRPGFRLLDDLVTCASRNPCSSSPCRGGATCVLGPHGKNYTCRCPQGYQLDSSQLDCVDVDECQDSPCAQECVNTPGGFRCECWVGYEPGGPGEGACQDVDECALGRSPCAQGCTNTDGSFHCSCEEGYVLAGEDGTQCQDVDECVGPGGPLCDSLCFNTQGSFHCGCLPGWVLAPNGVSCTMGPVSLGPPSGPPDEEDKGEKEGSTVPRAATASPTRGPEGTPKATPTTSRPSLSSDAPITSAPLKMLAPSGSPGVWREPSIHHATAASGPQEPAGGDSSVATQNNDGTDGQK
Specifications

Specifications

ABM85160.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
86.9kDa
12.5% SDS-PAGE Stained with Coomassie Blue
ADTEAVVCVGTACYTAHSGKLSAAEAQNHCNQNGGNLATVKSKEEAQHVQRVLAQLLRREAALTARMSKFWIGLQREKGKCLDPSLPLKGFSWVGGGEDTPYSNWHKELRNSCISKRCVSLLLDLSQPLLPSRLPKWSEGPCGSPGSPGSNIEGFVCKFSFKGMCRPLALGGPGQVTYTTPFQTTSSSLEAVPFASAANVACGEGDKDETQSHYFLCKEKAPDVFDWGSSGPLCVSPKYGCNFNNGGCHQDCFEGGDGSFLCGCRPGFRLLDDLVTCASRNPCSSSPCRGGATCVLGPHGKNYTCRCPQGYQLDSSQLDCVDVDECQDSPCAQECVNTPGGFRCECWVGYEPGGPGEGACQDVDECALGRSPCAQGCTNTDGSFHCSCEEGYVLAGEDGTQCQDVDECVGPGGPLCDSLCFNTQGSFHCGCLPGWVLAPNGVSCTMGPVSLGPPSGPPDEEDKGEKEGSTVPRAATASPTRGPEGTPKATPTTSRPSLSSDAPITSAPLKMLAPSGSPGVWREPSIHHATAASGPQEPAGGDSSVATQNNDGTDGQK
RUO
CD93
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
22918
CD93 (Human) Recombinant Protein (Q03)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1QR1/C1qR(P)/C1qRP/CDw93/MXRA4/dJ737E23.1
CD93
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers