missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD83 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2044
This item is not returnable.
View return policy
Description
CD83 Polyclonal specifically detects CD83 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| CD83 | |
| Polyclonal | |
| Unconjugated | |
| Q01151 | |
| CD83 | |
| Mouse myeloma cell line NS0-derived recombinant human CD83 Thr20-Ala143 Accession # Q01151 | |
| 100 μg | |
| Primary | |
| Detects human CD83 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse CD83 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| B-cell activation protein, BL11, BL11CD83 antigen, CD83, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15, HB15hCD83 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9308 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction