missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD81 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£205.00 - £476.00
Specifications
| Antigen | CD81 |
|---|---|
| Clone | 2664C |
| Dilution | Western Blot 4 ug/mL, Simple Western 10 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18790887
![]() |
R&D Systems
MAB46152-SP |
25 μg |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760447
![]() |
R&D Systems
MAB46152-100 |
100 μg |
£476.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD81 Monoclonal antibody specifically detects CD81 in Human samples. It is validated for Western Blot, Western BlotSpecifications
| CD81 | |
| Western Blot 4 ug/mL, Simple Western 10 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD81, CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, TSPAN28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| CHO-derived human CD81 protein, Phe113-Ala201, Accession # P60033 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2664C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P60033 | |
| 975 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title