missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD79B Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6620-SP
This item is not returnable.
View return policy
Description
CD79B Polyclonal specifically detects CD79B in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| CD79B | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B29, B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79B, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGB, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 974 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P40259 | |
| CD79B | |
| Chinese hamster cell line CHO-derived recombinant human CD79B Ala29-Asp159 Accession # P40259 | |
| 25 μg | |
| Primary | |
| Detects human CD79B in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse CD79B is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction