missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD5L ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human CD5L quantitates human CD5L in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD5L.
CD5L, also known as CD5 antigen-like, is a protein that is expressed on the surface of various immune cells, including T cells, B cells, and natural killer cells. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including autoimmune disorders, cancer, and infectious diseases. CD5L is known to modulate T cell activation and cytokine production, and has been shown to have anti-inflammatory effects.
Specifications
Specifications
| Accession Number | 1/6; AAC-11; AI047839; AIM; AIM/Spalpha; API6; apoptosis inhibitor 6; apoptosis inhibitor expressed by macrophages; apoptosis inhibitor of macrophage; Apoptosis inhibitory 6; CD5 antigen-like; CD5 antigen-like (scavenger receptor cysteine rich family); CD5 molecule like; Cd5 molecule-like; Cd5l; CT2; CT-2; hAIM; IgM-associated peptide; mAIM; Pdp; Pdp 1/6; PRO229; Spalpha; SP-ALPHA; UNQ203/PRO229 |
| Assay Range | 0.5 to 120 pg/mL |
| Assay Sensitivity | 0.5 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 922 |
| Gene Symbol | Cd5l |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction