missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD55/DAF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2009
This item is not returnable.
View return policy
Description
CD55/DAF Monoclonal specifically detects CD55/DAF in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| CD55/DAF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.5-2 ug/mL | |
| P08174.4 | |
| CD55 | |
| NS0-derived recombinant human CD55/DAF Asp35-Ser353 Accession # P08174.4 | |
| 500 μg | |
| Primary | |
| Detects human CD55/DAF in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD97 is observed. | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| 278803 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD55, CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CR, CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, DAF, decay accelerating factor for complement, TC | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1604 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction