missing translation for 'onlineSavingsMsg'
Learn More

Abcam Human CD47 ELISA Kit - Extracellular

Product Code. 32114964 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 384 Tests
1 x 96 Tests
10 x 96 Tests
Unit Size:
384 tests
96 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32114964 10 x 96 Tests 96 tests
32111724 1 x 96 Tests 96 tests
32113468 1 x 384 Tests 384 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32114964 Supplier Abcam Supplier No. ab32440910x96Tests
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human CD47 ELISA Kit - Extracellular

Human CD47 ELISA Kit - Extracellular

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Accession Number Q08722
Assay Range 187.5 to 3000 pg/mL
Assay Sensitivity 46.6 pg/mL
Content And Storage 4°C
Detectable Analytes CD47
Assay Human CD47 ELISA Kit - Extracellular
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Sample Type Cell Culture Media, Plasma (Citrate), Plasma (EDTA), Plasma (Heparin), Serum
Sufficient For 10 x 96 Tests
For Use With (Application) ELISA
Gene CD47
Immunoassay Kit Grade RUO
Measuring Range 187.5 to 3000 pg/mL
Protein CD47
Quantitation Range 187.5 to 3000 pg/mL
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Immuno-oncology
Shipping Condition Blue Ice
Target CD47
Target Kit Named CD47
UniProt ID Q08722
Detection Limit 187.5 pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.