missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD45 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£169.00 - £415.00
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 1043745 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773717
![]() |
R&D Systems
MAB14304-SP |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773467
![]() |
R&D Systems
MAB14304-100 |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for ELISASpecifications
| CD45 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen | |
| Mouse myeloma cell line NS0-derived human CD45 protein, Gln24-Lys575, Accession # P08575 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1043745 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08575 | |
| 5788 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title