missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD45 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£181.00 - £393.00
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 998215 |
| Dilution | Immunohistochemistry 5-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793907
![]() |
R&D Systems
MAB14302-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18753077
![]() |
R&D Systems
MAB14302-100 |
100 μg |
£393.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45 Monoclonal specifically detects CD45 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
| CD45 | |
| Immunohistochemistry 5-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Mouse myeloma cell line NS0-derived recombinant human CD45, Thr24-Asn575, Accession No. P08575 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD45 in direct ELISA. |
| 998215 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Human | |
| P08575 | |
| 5788.0 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title