missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD44s Pan Specific Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 22 publications
Brand: R&D Systems BBA10
This item is not returnable.
View return policy
Description
CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.
Specifications
| CD44 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation, CyTOF-ready | |
| CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 960 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF | |
| 2C5 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| CD44 | |
| Recombinant human CD44v3-10 (includes the invariant N-terminal exons and CD44v3-10 exons) | |
| 200 μg | |
| Primary | |
| Detects human CD44s on a panel of CD44 transfected COS cells by flow cytometry (Fox, S.B. et al. (1994) Cancer Res. 54:4539). This antibody recognizes an epitope in the invariant N-terminal region of all CD44 protein isoforms. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction