missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD44 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£185.00 - £582.00
Specifications
| Antigen | CD44 |
|---|---|
| Clone | 1058722 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18764637
![]() |
R&D Systems
MAB10146-SP |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792817
![]() |
R&D Systems
MAB10146-100 |
100 μg |
£582.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD44 Monoclonal antibody specifically detects CD44 in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| CD44 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Chinese Hamster Ovary cell line, CHO-derived human CD44, Gln21-Thr222, Accession # P16070 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1058722 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P16070 | |
| 960 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title