missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD44 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF3660-SP
This item is not returnable.
View return policy
Description
CD44 Polyclonal specifically detects CD44 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| CD44 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 960 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P16070 | |
| CD44 | |
| Mouse myeloma cell line NS0-derived recombinant human CD44 Gln21-Pro220 Accession # P16070 | |
| 25 μg | |
| Primary | |
| Detects human CD44 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse CD44 is observed and approximately 12% cross-reactivity with recombinant rat CD44 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction