missing translation for 'onlineSavingsMsg'
Learn More

Human CD39/ENTPD1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18756078
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18756078 25 μg 25µg
18142901 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18756078 Supplier R&D Systems Supplier No. MAB43971SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39/ENTPD1
Applications Western Blot
Classification Monoclonal
Clone 498409
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P49961
Gene Alias ADPase, ATPDase, ATP-diphosphatase, CD39, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, ENTPD1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPD1, NTPDase 1, NTPDase-1
Gene Symbols ENTPD1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CD39 Thr38-Val478 Accession # P49961
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 953
Test Specificity Detects human CD39/ENTPD1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse CD39 or recombinant human CD39L2, 3, or 4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.