missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD39/ENTPD1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB43971
This item is not returnable.
View return policy
Description
CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Human samples. It is validated for Western Blot.
Specifications
| CD39/ENTPD1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P49961 | |
| ENTPD1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD39 Thr38-Val478 Accession # P49961 | |
| 100 μg | |
| Primary | |
| Detects human CD39/ENTPD1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse CD39 or recombinant human CD39L2, 3, or 4 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 498409 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADPase, ATPDase, ATP-diphosphatase, CD39, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, ENTPD1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPD1, NTPDase 1, NTPDase-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 953 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction