missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD38 (Research Grade Daratumumab Biosimilar) Antibody, Novus Biologicals™

Human Monoclonal Antibody
Brand: R&D Systems MAB11493-100
This item is not returnable.
View return policy
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for ELISA
Specifications
| CD38 | |
| Monoclonal | |
| Unconjugated | |
| P28907 | |
| Human | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 952 | |
| Human | |
| Purified |
| ELISA | |
| Hu208 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl Cyclase, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose Hydrolase, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 | |
| NS0 mouse myeloma cell line transfected with human CD38, Met1-Ile300, Accession # P28907 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction