missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human CD35 - Ready-To-Use ELISA Kit (Colorimetric)

Product Code. 30056527
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30056527 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30056527 Supplier Novus Biologicals™ Supplier No. NBP331283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.312 - 20 ng/mL (example only; lot dependent)
Assay Sensitivity 0.11 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Tissue Homogenates, Biological Fluids
For Use With (Application) ELISA
Gene ID (Entrez) 1378
Gene Symbol CR1
Quantity 1 Kit
Research Discipline Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym complement component (3b/4b) receptor 1 (Knops blood group), KN
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.