missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD31/PECAM-1 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£206.00 - £476.00
Specifications
| Antigen | CD31/PECAM-1 |
|---|---|
| Clone | 1094431 |
| Dilution | Western Blot 2 μg/mL, Simple Western 5 μg/mL, Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL |
| Applications | Western Blot, Simple Western, Immunohistochemistry, Multiplex Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306746
|
R&D Systems
MAB11666-SP |
25 μg |
£206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306751
|
R&D Systems
MAB11666-100 |
100 μg |
£476.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry,Multiplex ImmunofluorescenceSpecifications
| CD31/PECAM-1 | |
| Western Blot 2 μg/mL, Simple Western 5 μg/mL, Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human CD31/PECAM01, Gln28-Lys601, Accession # P16284 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Antibody |
| 1094431 | |
| Western Blot, Simple Western, Immunohistochemistry, Multiplex Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| P16284 | |
| 5175 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title