missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD302/CLEC13A Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6367
This item is not returnable.
View return policy
Description
CD302/CLEC13A Polyclonal specifically detects CD302/CLEC13A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| CD302/CLEC13A | |
| Polyclonal | |
| Unconjugated | |
| Q8IX05 | |
| CD302 | |
| Mouse myeloma cell line NS0-derived human CD302/CLEC13A Ala20-Asn167 Accession # Q8IX05 | |
| 100 μg | |
| Primary | |
| Detects human CD302/CLEC13A in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse CD302 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| BIMLEC, CD302, CD302 antigenC-type lectin domain family 13, member A, CD302 molecule, CLEC13A, CLEC13AMGC22301, C-type lectin domain family 13 member A, DCL1, DCL-1, DCL1C-type lectin BIMLEC, KIAA0022FLJ43091, Type I transmembrane C-type lectin receptor DCL-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9936 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction